Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177874.1 | NP_001171345.1 | 2 - 3 | 164 | 65 | |
NM_145370.2 | NP_663345.2 | 2 - 3 | 304 | 65 | ||
GenBank mRNA | AK088490.1 | - | 2 - 3 | 161 | 65 | |
AK190400.1 | - | 1 - 2 | 47 | 65 | ||
BC003350.1 | - | 2 - 3 | 304 | 3481979 | 65 | |
BC103782.1 | - | 2 - 3 | 131 | 6486599 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |