Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145537.2 | NP_663512.2 | 6 - 7 | 835 | 75 | |
GenBank mRNA | AK029164.1 | - | 6 - 7 | 787 | 75 | |
AK075650.1 | - | 6 - 7 | 749 | 75 | ||
AK079296.1 | - | 6 - 7 | 771 | 75 | ||
AK080636.1 | - | 6 - 7 | 780 | 75 | ||
AK149892.1 | - | 6 - 7 | 771 | 75 | ||
AK155024.1 | - | 6 - 7 | 798 | 75 | ||
AK166998.1 | - | 6 - 7 | 771 | 75 | ||
AK183727.1 | - | 1 - 2 | 224 | 75 | ||
BC008268.1 | - | 6 - 7 | 746 | 2811074 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |