Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144786.2 | NP_659035.2 | 9 - 10 | 1285 | 61 | |
GenBank mRNA | AK032051.1 | - | 9 - 10 | 1283 | 61 | |
AK131175.1 | - | 6 - 7 | 3308 | 61 | ||
BC005772.1 | - | 9 - 10 | 1266 | 3602314 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |