Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001144.3 | NP_001001144.2 | 21 - 22 | 3626 | 95 | |
NM_001103162.2 | NP_001096632.1 | 21 - 22 | 3538 | 95 | ||
GenBank mRNA | AK035432.1 | - | 21 - 22 | 3517 | 95 | |
AK146658.1 | - | 21 - 22 | 3608 | 95 | ||
AK150228.1 | - | 20 - 21 | 3542 | 95 | ||
AK150275.1 | - | 21 - 22 | 3510 | 95 | ||
AK152478.1 | - | 21 - 22 | 3510 | 95 | ||
AK153330.1 | - | 21 - 22 | 3510 | 95 | ||
AK172912.1 | - | 19 - 20 | 4386 | 95 | ||
BC039638.1 | - | 6 - 7 | 978 | 95 | ||
BC051066.1 | - | 7 - 8 | 1190 | 95 | ||
BC055472.1 | - | 2 - 3 | 165 | 95 | ||
BC069955.1 | - | 2 - 3 | 165 | 95 | ||
BC070437.1 | - | 21 - 22 | 3626 | 30535805 | 95 | |
BC072633.1 | - | 21 - 22 | 3499 | 6849461 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |