Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5rap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025876.2 | NP_080152.1 | 8 - 9 | 1156 | 144 | |
GenBank mRNA | AK010081.1 | - | 8 - 9 | 1156 | 144 | |
AK010097.1 | - | 8 - 9 | 1156 | 144 | ||
AK088517.1 | - | 8 - 9 | 1165 | 144 | ||
BC025132.1 | - | 7 - 8 | 1121 | 3666341 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |