Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016776.2 | NP_058056.2 | 14 - 15 | 1917 | 66 | |
GenBank mRNA | AK083908.1 | - | 14 - 15 | 1943 | 66 | |
AK143469.1 | - | 14 - 15 | 1942 | 66 | ||
AK143487.1 | - | 14 - 15 | 1950 | 66 | ||
AK149747.1 | - | 9 - 10 | 1226 | 66 | ||
AK154016.1 | - | 12 - 13 | 2136 | 66 | ||
AK155074.1 | - | 14 - 15 | 1928 | 66 | ||
BC004078.1 | - | 4 - 5 | 401 | 66 | ||
BC015070.1 | - | 14 - 15 | 1918 | 66 | ||
BC048858.1 | - | 14 - 15 | 1917 | 5363740 | 66 | |
BC052889.1 | - | 14 - 15 | 1938 | 30057169 | 66 | |
BC060167.1 | - | 14 - 15 | 1923 | 66 | ||
U63648.1 | - | 14 - 15 | 1926 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |