Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164369.1 | NP_001157841.1 | 7 - 8 | 1238 | 66 | |
NM_029815.2 | NP_084091.2 | 8 - 9 | 1376 | 66 | ||
GenBank mRNA | AK008957.1 | - | 5 - 6 | 782 | 66 | |
AY219233.1 | - | 8 - 9 | 1359 | 66 | ||
BC113166.1 | - | 7 - 8 | 1218 | 40059981 | 66 | |
BC113957.1 | - | 7 - 8 | 1218 | 40059979 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |