Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009981.2 | NP_001009981.1 | 8 - 9 | 940 | 68 | |
NM_001282109.1 | NP_001269038.1 | 8 - 9 | 948 | 68 | ||
NM_029736.2 | NP_084012.1 | 9 - 10 | 1024 | 68 | ||
GenBank mRNA | AK010834.1 | - | 9 - 10 | 974 | 68 | |
AK040517.1 | - | 9 - 10 | 903 | 68 | ||
AK152752.1 | - | 9 - 10 | 985 | 68 | ||
AK152849.1 | - | 9 - 10 | 985 | 68 | ||
AK160402.1 | - | 9 - 10 | 1008 | 68 | ||
AK165983.1 | - | 8 - 9 | 885 | 68 | ||
AY825926.1 | - | 9 - 10 | 777 | 68 | ||
AY825928.1 | - | 8 - 9 | 693 | 68 | ||
BC116666.1 | - | 9 - 10 | 890 | 40107077 | 68 | |
BC116717.1 | - | 9 - 10 | 890 | 40107078 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |