Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029876.1 | NP_001025047.1 | 1 - 2 | 120 | 67 | |
GenBank mRNA | AK133906.1 | - | 2 - 3 | 434 | 67 | |
BC151125.1 | - | 1 - 2 | 199 | 9088038 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |