Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146254.4 | NP_666366.4 | 9 - 10 | 1514 | 100 | |
GenBank mRNA | AK030997.1 | - | 2 - 3 | 3199 | 100 | |
BC028975.1 | - | 9 - 10 | 1439 | 5363117 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |