Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039081.2 | NP_001034170.1 | 4 - 5 | 440 | 147 | |
NM_001286221.1 | NP_001273150.1 | 6 - 7 | 571 | 147 | ||
NM_001286222.1 | NP_001273151.1 | 6 - 7 | 625 | 147 | ||
NM_001286223.1 | NP_001273152.1 | 4 - 5 | 421 | 147 | ||
NM_027696.3 | NP_081972.2 | 6 - 7 | 628 | 147 | ||
GenBank mRNA | AK129405.1 | - | 6 - 7 | 548 | 147 | |
AK138876.1 | - | 4 - 5 | 441 | 147 | ||
AK143201.1 | - | 4 - 5 | 545 | 148 | ||
AK168693.1 | - | 4 - 5 | 421 | 147 | ||
AY769435.1 | - | 5 - 6 | 420 | 147 | ||
AY769436.1 | - | 4 - 5 | 343 | 147 | ||
AY769437.1 | - | 4 - 5 | 393 | 147 | ||
AY769438.1 | - | 6 - 7 | 493 | 147 | ||
BC043930.1 | - | 6 - 7 | 553 | 147 | ||
BC108273.1 | - | 4 - 5 | 407 | 30080243 | 147 | |
BC130227.1 | - | 4 - 5 | 392 | 40141863 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |