Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm7sf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026281.2 | NP_080557.2 | 3 - 4 | 508 | 64 | |
GenBank mRNA | AK010720.1 | - | 3 - 4 | 351 | 64 | |
AK154633.1 | - | 3 - 4 | 508 | 64 | ||
AK178685.1 | - | 1 - 2 | 85 | 64 | ||
AK195575.1 | - | 1 - 2 | 85 | 64 | ||
BC023389.1 | - | 3 - 4 | 446 | 64 | ||
DQ104329.1 | - | 3 - 4 | 456 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |