Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stradb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172656.5 | NP_766244.4 | 1 - 2 | 281 | 148 | |
GenBank mRNA | AB057666.1 | - | 1 - 2 | 48 | 148 | |
AB057667.1 | - | 1 - 2 | 48 | 148 | ||
AK035207.1 | - | 1 - 2 | 212 | 148 | ||
AK036678.1 | - | 1 - 2 | 276 | 148 | ||
AK046790.1 | - | 254 | 148 | |||
AK080096.1 | - | 1 - 2 | 245 | 148 | ||
AK086974.1 | - | 1 - 2 | 212 | 148 | ||
AK140272.1 | - | 1 - 2 | 331 | 148 | ||
BC037003.1 | - | 1 - 2 | 190 | 148 | ||
BC038492.1 | - | 1 - 2 | 280 | 148 | ||
BC055814.1 | - | 1 - 2 | 254 | 148 | ||
BC060956.1 | - | 1 - 2 | 138 | 30316511 | 148 | |
CJ060534.1 | - | 1 - 2 | 331 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |