Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033606.2 | NP_001028778.2 | 9 - 10 | 1606 | 74 | |
NM_001136222.1 | NP_001129694.1 | 8 - 9 | 1188 | 74 | ||
NM_028817.3 | NP_083093.2 | 10 - 11 | 1746 | 74 | ||
GenBank mRNA | AK012088.1 | - | 9 - 10 | 1549 | 74 | |
AK160839.1 | - | 8 - 9 | 1132 | 74 | ||
AK161708.1 | - | 10 - 11 | 1747 | 74 | ||
AK166723.1 | - | 10 - 11 | 1684 | 74 | ||
BC031529.1 | - | 8 - 9 | 1331 | 4009004 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |