Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278075.1 | NP_001265004.1 | 4 - 5 | 397 | 76 | |
NM_011811.4 | NP_035941.2 | 4 - 5 | 397 | 76 | ||
NR_102757.1 | 4 - 5 | 299 | 76 | |||
NR_102758.1 | 3 - 4 | 341 | 76 | |||
GenBank mRNA | AF123263.1 | - | 4 - 5 | 397 | 76 | |
AK010271.1 | - | 4 - 5 | 360 | 76 | ||
AK012154.1 | - | 4 - 5 | 360 | 76 | ||
AK045432.1 | - | 4 - 5 | 349 | 76 | ||
AK076607.1 | - | 4 - 5 | 359 | 76 | ||
AK134799.1 | - | 3 - 4 | 303 | 76 | ||
AK165645.1 | - | 4 - 5 | 264 | 76 | ||
AK167945.1 | - | 4 - 5 | 360 | 76 | ||
AK168923.1 | - | 4 - 5 | 341 | 76 | ||
AK169621.1 | - | 3 - 4 | 304 | 76 | ||
BC002313.1 | - | 4 - 5 | 256 | 3592805 | 76 | |
BC016428.1 | - | 4 - 5 | 364 | 4239906 | 76 | |
CN693684.1 | - | 1 - 2 | 85 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |