Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mysm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177239.2 | NP_796213.2 | 8 - 9 | 1273 | 105 | |
GenBank mRNA | AK173300.1 | - | 5 - 6 | 1015 | 105 | |
BC150946.1 | - | 8 - 9 | 1269 | 9087857 | 105 | |
BC151172.1 | - | 8 - 9 | 1269 | 9088061 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |