Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100004414.mm.1 |
NM_139270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004414.mm.1 |
NM_139270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004414.mm.1 |
TC0100000546.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100004417.mm.1 |
NM_139270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004417.mm.1 |
NM_139270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004417.mm.1 |
TC0100000546.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100004421.mm.1 |
NM_139270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004421.mm.1 |
NM_139270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004421.mm.1 |
TC0100000546.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100004422.mm.1 |
NM_139270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004422.mm.1 |
NM_139270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004422.mm.1 |
TC0100000546.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100004416.mm.1 |
NM_139270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004416.mm.1 |
NM_139270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004416.mm.1 |
TC0100000546.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100004415.mm.1 |
NM_139270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004415.mm.1 |
NM_139270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004415.mm.1 |
TC0100000546.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100004418.mm.1 |
NM_139270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004418.mm.1 |
NM_139270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004418.mm.1 |
TC0100000546.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100004419.mm.1 |
NM_139270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004419.mm.1 |
NM_139270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004419.mm.1 |
TC0100000546.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100004420.mm.1 |
NM_139270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004420.mm.1 |
NM_139270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004420.mm.1 |
TC0100000546.mm.1 |
MTA TC |