Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mad2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK009587.1 | - | 3 - 4 | 541 | 66 | |
AK150216.1 | - | 3 - 4 | 534 | 66 | ||
AK165627.1 | - | 3 - 4 | 345 | 66 | ||
BC011282.1 | - | 3 - 4 | 471 | 3484522 | 66 | |
BC071264.1 | - | 5 - 6 | 471 | 6516319 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |