Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK140734.1 | - | 11 - 12 | 1220 | 98 | |
AK220466.1 | - | 11 - 12 | 1229 | 98 | ||
BC021396.1 | - | 10 - 11 | 1259 | 98 | ||
BC070412.1 | - | 12 - 13 | 1394 | 5705112 | 98 | |
U96963.1 | - | 10 - 11 | 1232 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |