Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134134.2 | NP_598895.2 | 18 - 19 | 3455 | 63 | |
NM_178277.1 | NP_840061.1 | 17 - 18 | 3550 | 63 | ||
GenBank mRNA | AB045322.1 | - | 18 - 19 | 3456 | 63 | |
AK129085.1 | - | 14 - 15 | 2458 | 63 | ||
AK132529.1 | - | 11 - 12 | 1934 | 63 | ||
AK178113.1 | - | 2 - 3 | 313 | 63 | ||
BC025148.1 | - | 3 - 4 | 418 | 63 | ||
BC058089.1 | - | 15 - 16 | 2848 | 63 | ||
BC060046.1 | - | 11 - 12 | 1880 | 63 | ||
BC066029.1 | - | 12 - 13 | 2686 | 6417061 | 63 | |
BC079580.1 | - | 17 - 18 | 3498 | 5702202 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |