Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021541.3 | NP_067516.1 | 1 - 2 | 63 | 100 | |
GenBank mRNA | AJ272227.1 | - | 1 - 2 | 55 | 100 | |
CF740624.1 | - | 1 - 2 | 62 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus 1700 Probe spans exons |