Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281980.1 | NP_001268909.1 | 15 - 16 | 1289 | 65 | |
NM_001281981.1 | NP_001268910.1 | 16 - 17 | 1330 | 65 | ||
NM_008511.3 | NP_032537.3 | 15 - 16 | 1561 | 65 | ||
GenBank mRNA | AK153865.1 | - | 15 - 16 | 1291 | 65 | |
AK166635.1 | - | 13 - 14 | 1010 | 65 | ||
AK168775.1 | - | 13 - 14 | 987 | 65 | ||
AK169578.1 | - | 16 - 17 | 1314 | 65 | ||
AY753324.1 | - | 14 - 15 | 1124 | 65 | ||
BC052909.1 | - | 15 - 16 | 1497 | 30076553 | 65 | |
U10484.1 | - | 15 - 16 | 1267 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |