Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173774.3 | NP_776135.2 | 3 - 4 | 669 | 66 | |
GenBank mRNA | AK082651.1 | - | 661 | 66 | ||
AK134677.1 | - | 3 - 4 | 593 | 66 | ||
AK134683.1 | - | 3 - 4 | 659 | 66 | ||
CX241103.1 | - | 3 - 4 | 587 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mus musculus Probe spans exons |