Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144946.4 | NP_659195.3 | 1 - 2 | 959 | 133 | |
GenBank mRNA | AF448840.1 | - | 1 - 2 | 292 | 133 | |
AK043710.1 | - | 1 - 2 | 295 | 133 | ||
AK081325.1 | - | 1 - 2 | 960 | 133 | ||
BC051145.1 | - | 1 - 2 | 319 | 6590345 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons |