Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142950.1 | NP_001136422.1 | 13 - 14 | 1514 | 60 | |
NM_027350.3 | NP_081626.2 | 13 - 14 | 1511 | 60 | ||
GenBank mRNA | AK013880.1 | - | 4 - 5 | 491 | 60 | |
AK077699.1 | - | 13 - 14 | 1492 | 60 | ||
BC052849.1 | - | 13 - 14 | 1472 | 30057822 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |