Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037711.2 | NP_001032800.2 | 8 - 9 | 1752 | 76 | |
NM_001293727.1 | NP_001280656.1 | 8 - 9 | 1728 | 76 | ||
GenBank mRNA | AK173148.1 | - | 8 - 9 | 1729 | 76 | |
BC042459.1 | - | 8 - 9 | 1698 | 76 | ||
BC052941.1 | - | 8 - 9 | 1674 | 30021945 | 76 | |
BC064474.1 | - | 8 - 9 | 1676 | 76 | ||
BC108378.1 | - | 30736765 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |