Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013783.2 | NP_038811.2 | 5 - 6 | 745 | 58 | |
GenBank mRNA | AF157105.1 | - | 5 - 6 | 691 | 58 | |
AF157106.1 | - | 3 - 4 | 400 | 58 | ||
AF176569.1 | - | 5 - 6 | 742 | 58 | ||
AF302075.1 | - | 3 - 4 | 366 | 58 | ||
AF302076.1 | - | 4 - 5 | 435 | 58 | ||
AF302077.1 | - | 3 - 4 | 366 | 58 | ||
AK154366.1 | - | 2 - 3 | 234 | 58 | ||
BC137649.1 | - | 3 - 4 | 518 | 8860668 | 58 | |
BC144804.1 | - | 3 - 4 | 518 | 9053352 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |