Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053255.3 | NP_444485.2 | 1 - 2 | 75 | 110 | |
GenBank mRNA | AF308697.1 | - | 1 - 2 | 74 | 109 | |
BY286576.1 | - | 1 - 2 | 76 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |