Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015798.3 | NP_056613.1 | 6 - 7 | 949 | 114 | |
GenBank mRNA | AF176530.1 | - | 6 - 7 | 933 | 114 | |
AK082809.1 | - | 6 - 7 | 970 | 114 | ||
AK163224.1 | - | 6 - 7 | 950 | 114 | ||
AK166591.1 | - | 6 - 7 | 950 | 114 | ||
AK166599.1 | - | 6 - 7 | 951 | 114 | ||
AK166918.1 | - | 6 - 7 | 946 | 114 | ||
AK167118.1 | - | 6 - 7 | 950 | 114 | ||
AK167270.1 | - | 6 - 7 | 949 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |