Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008891.2 | NP_032917.2 | 6 - 7 | 578 | 128 | |
GenBank mRNA | AK009055.1 | - | 6 - 7 | 533 | 128 | |
AK028219.1 | - | 6 - 7 | 519 | 128 | ||
AK030978.1 | - | 6 - 7 | 507 | 128 | ||
AK050669.1 | - | 6 - 7 | 524 | 128 | ||
AK157587.1 | - | 6 - 7 | 522 | 128 | ||
AK160616.1 | - | 6 - 7 | 513 | 128 | ||
AK161109.1 | - | 6 - 7 | 518 | 128 | ||
BC132571.1 | - | 6 - 7 | 512 | 40130848 | 128 | |
BC138014.1 | - | 6 - 7 | 512 | 8861033 | 128 | |
Y08701.1 | - | 6 - 7 | 509 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |