Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168491.1 | NP_001161963.1 | 7 - 8 | 1076 | 110 | |
NM_001168492.1 | NP_001161964.1 | 7 - 8 | 1076 | 110 | ||
NM_011050.4 | NP_035180.2 | 7 - 8 | 1164 | 110 | ||
GenBank mRNA | AK134366.1 | - | 7 - 8 | 1165 | 110 | |
AK172654.1 | - | 7 - 8 | 1032 | 110 | ||
BC055739.1 | - | 7 - 8 | 1011 | 6414089 | 110 | |
CT010188.1 | - | 6 - 7 | 874 | 110 | ||
D50465.1 | - | 6 - 7 | 944 | 110 | ||
D86344.1 | - | 7 - 8 | 960 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |