Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010123.3 | NP_034253.3 | 13 - 14 | 2264 | 118 | |
GenBank mRNA | AK013070.1 | - | 3 - 4 | 579 | 118 | |
AK145629.1 | - | 13 - 14 | 2229 | 118 | ||
AK162784.1 | - | 2 - 3 | 177 | 118 | ||
AK167010.1 | - | 13 - 14 | 2231 | 118 | ||
AK172899.1 | - | 13 - 14 | 2201 | 118 | ||
U14172.1 | - | 13 - 14 | 2214 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |