Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033621.3 | NP_001028793.1 | 1 - 2 | 82 | 82 | |
GenBank mRNA | AF230979.2 | - | 1 - 2 | 82 | 82 | |
AK017447.1 | - | 1 - 2 | 83 | 82 | ||
BC016214.1 | - | 1 - 2 | 60 | 4482382 | 82 | |
BC064052.1 | - | 1 - 2 | 55 | 1366418 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |