Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163732.1 | NP_001157204.1 | 11 - 12 | 1164 | 81 | |
NM_172869.4 | NP_766457.1 | 11 - 12 | 1164 | 81 | ||
GenBank mRNA | AK034956.1 | - | 11 - 12 | 1139 | 81 | |
AK048561.1 | - | 11 - 12 | 1107 | 81 | ||
AK079349.1 | - | 11 - 12 | 1168 | 81 | ||
BC037727.1 | - | 11 - 12 | 1335 | 81 | ||
BC112419.1 | - | 11 - 12 | 1072 | 40061426 | 81 | |
BC113795.1 | - | 11 - 12 | 1072 | 40061428 | 81 | |
BC118964.2 | - | 11 - 12 | 1074 | 40097059 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |