Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027326.3 | NP_081602.3 | 9 - 10 | 1815 | 114 | |
NM_029931.3 | NP_084207.2 | 5 - 6 | 488 | 114 | ||
GenBank mRNA | AF333960.1 | - | 8 - 9 | 1476 | 114 | |
AK008707.1 | - | 5 - 6 | 450 | 114 | ||
AK136066.1 | - | 5 - 6 | 1049 | 114 | ||
AK147009.1 | - | 5 - 6 | 402 | 114 | ||
BC021420.1 | - | 10 - 11 | 1654 | 5012671 | 114 | |
BU614509.1 | - | 5 - 6 | 616 | 114 | ||
DV051568.1 | - | 498 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |