Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290993.1 | NP_001277922.1 | 4 - 5 | 1104 | 145 | |
NM_011774.3 | NP_035904.2 | 5 - 6 | 1251 | 145 | ||
GenBank mRNA | AF003747.1 | - | 5 - 6 | 1052 | 145 | |
AK134245.1 | - | 5 - 6 | 145 | |||
AK166106.1 | - | 4 - 5 | 1106 | 145 | ||
BC117997.1 | - | 5 - 6 | 1109 | 40095367 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |