Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013767.2 | NP_001013789.1 | 10 - 11 | 1220 | 113 | |
GenBank mRNA | AY578330.1 | - | 10 - 11 | 1220 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Mus musculus Probe spans exons |