Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198167.3 | NP_937810.2 | 8 - 9 | 724 | 74 | |
GenBank mRNA | AK090200.1 | - | 8 - 9 | 645 | 74 | |
AK159673.1 | - | 8 - 9 | 729 | 74 | ||
BC059283.1 | - | 1 - 2 | 72 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |