Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128103.2 | NP_001121575.1 | 8 - 9 | 1016 | 72 | |
GenBank mRNA | AK045930.1 | - | 8 - 9 | 1017 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |