Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038660.2 | NP_001033749.1 | 1 - 2 | 779 | 67 | |
GenBank mRNA | AY795939.1 | - | 1 - 2 | 758 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mus musculus Probe spans exons |