Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110513.1 | NP_001103983.1 | 12 - 13 | 1247 | 62 | |
GenBank mRNA | AF387631.1 | - | 12 - 13 | 1247 | 62 | |
AF387632.1 | - | 12 - 13 | 1247 | 62 | ||
AF387633.1 | - | 12 - 13 | 1247 | 62 | ||
AF387634.1 | - | 12 - 13 | 1247 | 62 | ||
BC138119.1 | - | 12 - 13 | 1247 | 8861138 | 62 | |
BC145155.1 | - | 13 - 14 | 1333 | 9053706 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |