Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110145.1 | NP_001103615.1 | 6 - 7 | 730 | 89 | |
NM_009278.4 | NP_033304.1 | 6 - 7 | 757 | 89 | ||
GenBank mRNA | AK032940.1 | - | 6 - 7 | 674 | 89 | |
AK088650.1 | - | 6 - 7 | 713 | 89 | ||
AK088677.1 | - | 6 - 7 | 664 | 89 | ||
AK134864.1 | - | 6 - 7 | 695 | 89 | ||
AK141487.1 | - | 6 - 7 | 652 | 89 | ||
AK168828.1 | - | 6 - 7 | 680 | 89 | ||
BC003820.1 | - | 6 - 7 | 617 | 3593742 | 89 | |
BC106150.1 | - | 6 - 7 | 610 | 3483153 | 89 | |
L00993.1 | - | 6 - 7 | 693 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |