Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-T23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010398.3 | NP_034528.1 | 3 - 4 | 639 | 55 | |
GenBank mRNA | AB359230.2 | - | 3 - 4 | 639 | 55 | |
AF057279.2 | - | 607 | 55 | |||
AK133502.1 | - | 3 - 4 | 639 | 55 | ||
M11284.1 | - | 640 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |