Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcne3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190870.1 | NP_001177799.1 | 3 - 4 | 602 | 121 | |
GenBank mRNA | DQ832280.1 | - | 3 - 4 | 588 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |