Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020575.2 | NP_065600.1 | 3 - 4 | 506 | 61 | |
GenBank mRNA | AF155739.1 | - | 3 - 4 | 506 | 61 | |
AK034109.1 | - | 531 | 61 | |||
AK143177.1 | - | 3 - 4 | 526 | 61 | ||
AK166666.1 | - | 3 - 4 | 513 | 61 | ||
AK167246.1 | - | 3 - 4 | 488 | 61 | ||
AK168555.1 | - | 3 - 4 | 509 | 61 | ||
BC025029.1 | - | 3 - 4 | 470 | 4935124 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |