Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008027.2 | NP_032053.1 | 10 - 11 | 1111 | 62 | |
GenBank mRNA | AF145044.1 | - | 9 - 10 | 952 | 62 | |
AK150692.1 | - | 10 - 11 | 1133 | 62 | ||
AK150827.1 | - | 10 - 11 | 1060 | 62 | ||
AK167323.1 | - | 10 - 11 | 1038 | 62 | ||
AY167925.1 | - | 10 - 11 | 1117 | 62 | ||
BC004647.1 | - | 10 - 11 | 1121 | 3158071 | 62 | |
U90435.1 | - | 10 - 11 | 1111 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |