Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015754.2 | NP_056569.2 | 1 - 2 | 183 | 83 | |
GenBank mRNA | AK017606.1 | - | 1 - 2 | 124 | 83 | |
AK076944.1 | - | 1 - 2 | 178 | 83 | ||
BC011107.1 | - | 1 - 2 | 121 | 3982875 | 83 | |
BC050201.1 | - | 1 - 2 | 119 | 30013927 | 83 | |
BC147814.1 | - | 1 - 2 | 176 | 9087531 | 83 | |
BC147815.1 | - | 1 - 2 | 176 | 9087532 | 83 | |
BY256564.1 | - | 1 - 2 | 194 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |