Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172255.3 | NP_758459.2 | 4 - 5 | 646 | 105 | |
GenBank mRNA | AK043051.1 | - | 2 - 3 | 264 | 105 | |
AK166369.1 | - | 4 - 5 | 601 | 105 | ||
AK170058.1 | - | 4 - 5 | 594 | 105 | ||
BC037177.1 | - | 4 - 5 | 583 | 4948388 | 105 | |
CK638976.1 | - | 4 - 5 | 635 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |