Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178755.3 | NP_848870.2 | 15 - 16 | 2377 | 111 | |
GenBank mRNA | AK029744.1 | - | 15 - 16 | 2410 | 111 | |
AK029932.1 | - | 15 - 16 | 2378 | 111 | ||
AK030138.1 | - | 15 - 16 | 2365 | 111 | ||
AK030182.1 | - | 15 - 16 | 2424 | 111 | ||
AK041286.1 | - | 7 - 8 | 786 | 111 | ||
DQ867026.1 | - | 15 - 16 | 2410 | 111 | ||
DQ867027.1 | - | 15 - 16 | 2410 | 111 | ||
DQ867028.1 | - | 15 - 16 | 2410 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |