Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177785.1 | NP_001171256.1 | 5 - 6 | 990 | 79 | |
GenBank mRNA | X66084.1 | - | 5 - 6 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |